Tungsten ≥99.95%, rods, Ø 1.5 mm (0.06 in)

Supplier: Thermo Scientific Chemicals

42233-9X
CAAA42233-9XEA 113.29 CAD
CAAA42233-9X
Tungsten ≥99.95%, rods, Ø 1.5 mm (0.06 in)
Tungsten

~3.4g/10cm

Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR