2-Hydroxyacetophenone 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-1-phenylethanone, Benzoylcarbinol, Phenacyl hydroxyide, 2-Hydroxyacetophenone, Phenacyl alcohol, α-Hydroxyacetophenone

42013-06
CAAA42013-06EA 127.66 CAD
CAAA42013-06
2-Hydroxyacetophenone 97+%
2-Hydroxyacetophenone

MDL: MFCD00041829


Purity: 97+% CAS No.: 582-24-1 Molecular Formula: C8H8O2 Formula Weight: 136.15 Physical Form: Powder, Packaged under argon Sensitivity: Air and Moisture Sensitive MDL No.: MFCD00041829

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 118…120 °C (11 mmHg)
Melting Pt: 86…88 °C
Density: 1.0963 g/cm³ (99 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041829
CAS Number: 582-24-1
EINECS: 209-480-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR