Magnesium citrate hydrate (∼ 16% Mg) (dried basis) (dry weight)

Supplier: Thermo Scientific Chemicals

41921-A1 41921-A7
CAAAA41921-B5EA 1786.03 CAD
CAAAA41921-B5 CAAA41921-A1 CAAA41921-A7
Magnesium citrate hydrate (∼ 16% Mg) (dried basis) (dry weight)
Magnesium citrate hydrate

MDL: MFCD00036422


CAS No.: 3344-18-1 Molecular Formula: C12H10Mg3O14.xH2O Formula Weight: 451.13(anhy) Physical Form: Powder Appearance: White to off-white Merck Reference: 14,5663 MDL No.: MFCD09953657

Formula: C₁₂H₁₀Mg₃O₁₄·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD09953657
CAS Number: 3344-18-1
EINECS: 222-093-9
Merck Index: 14,5663

Order Now

Specification Test Results

Form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR