Gallium(II) telluride ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

41881-03 41881-06
CAAA41881-03EA 173.8 CAD
CAAA41881-03 CAAA41881-06
Gallium(II) telluride ≥99.999% (metals basis)
Gallium (II) telluride

MDL: MFCD00135536


Purity: 99.999% CAS No.: 12024-14-5 Molecular Formula: GaTe Formula Weight: 197.32 Physical Form: 6mm & down Sensitivity: Moisture Sensitive MDL No.: MFCD00135536

Formula: GaTe
MW: 197.32
Boiling Pt: 103…104 °C (0.08 mmHg)
Melting Pt: 824
Density: 5.44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00135536
CAS Number: 12024-14-5
EINECS: 234-690-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR