Sodium, Ion chromatography standard solution, Specpure®, Na|+ 1000 mg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

41696-AE
CAAA41696-AEEA 211.72 CAD
CAAA41696-AE
Sodium, Ion chromatography standard solution, Specpure®, Na|+ 1000 mg/ml, Thermo Scientific
Standards Cationic Standards Sodium Standards

Molecular Formula: NaCl in H2O
Physical Form: Liquid
MDL No.: MFCD00044631

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR