Styrene-4-sulfonic acid sodium salt

Supplier: Thermo Scientific Chemicals
Warning

41690-18 41690-30
CAAA41690-18EA 89.27 CAD
CAAA41690-18 CAAA41690-30
Styrene-4-sulfonic acid sodium salt
Styrene-4-sulfonic acid sodium salt

CAS No.: 2695-37-6 Molecular Formula: C8H7NaO3S Formula Weight: 206.19 Physical Form: Powder Appearance: White Beilstein Reference: 3575931 MDL No.: MFCD00013379

Formula: C₈H₇NaO₃S
Storage Temperature: Ambient
MDL Number: MFCD00013379
CAS Number: 2695-37-6
EINECS: 220-266-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR