Ytterbium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger

41423-09 41423-18
CAAA41423-09EA 221.2 CAD
CAAA41423-09 CAAA41423-18
Ytterbium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Ytterbium (III) fluoride

For vacuum deposition


Purity: 99.9% CAS No.: 13760-80-0 Molecular Formula: YbF3 Formula Weight: 230.04 Physical Form: 3-12mm Pieces Density: 8.2 Melting Point: 1157° UN No.: 3288 MDL No.: MFCD00049615


For vacuum deposition

Formula: F₃Yb
Boiling Pt: 2227 °C (1013 hPa)
Melting Pt: 1157 °C
Density: 8.2
Storage Temperature: Ambient
MDL Number: MFCD00049615
CAS Number: 13760-80-0
EINECS: 237-354-2
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR