Polyvinyl alcohol, M.W. ∼ 57,000-66,000 98-99, molecular weight_medium, hydrolysed

Supplier: Thermo Scientific Chemicals

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

41242-14 41242-22 41242-36
CAAA41242-14EA 51.19 CAD
CAAA41242-14 CAAA41242-22 CAAA41242-36
Polyvinyl alcohol, M.W. ∼ 57,000-66,000 98-99, molecular weight_medium, hydrolysed
Polyvinyl alcohol

Average M.W. 57,000-66,000


Purity: 98-99% CAS No.: 9002-89-5 Molecular Formula: [-CH2CH(OH)-]n Odor: Odorless MDL No.: MFCD00081922

Formula: [-CH₂CH(OH)-]n
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR