Chromium disilicide ≥99.5% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), thickness 6.35 mm (0.250 in)

Supplier: Thermo Scientific Chemicals
Danger

41063-KS
CAAA41063-KSEA 1572.1 CAD
CAAA41063-KS
Chromium disilicide ≥99.5% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), thickness 6.35 mm (0.250 in)
Chromium disilicide

Purity: 99.5% Molecular Formula: CrSi2 Formula Weight: 108.17 MDL No.: MFCD00168070

Formula: CrSi₂
Storage Temperature: Ambient
MDL Number: MFCD00168070
CAS Number: 12018-09-6
EINECS: 234-633-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR