Hastelloy C® alloy, gauze woven from wire, Hastelloy C® 20 mesh woven from 0.23mm (0.009in)

Supplier: Thermo Scientific Chemicals
Danger

40938-FL 40938-GT 40938-HB
CAAA40938-FLEA 64.15 CAD
CAAA40938-FL CAAA40938-GT CAAA40938-HB
Hastelloy C® alloy, gauze woven from wire, Hastelloy C® 20 mesh woven from 0.23mm (0.009in)
Hastelloy C® alloy

Applications: High strength, corrosion resistant alloy


Physical Form: Wire Cloth MDL No.: MFCD00801065


Applications: High strength, corrosion resistant alloy

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR