Aluminium ≥99.99% (metals basis), wire, hard, Ø 0.05 mm (0.002 in)

Supplier: Thermo Scientific Chemicals

40676-G5
CAAA40676-G1EA 407.17 CAD
CAAA40676-G1 CAAA40676-G5
Aluminium ≥99.99% (metals basis), wire, hard, Ø 0.05 mm (0.002 in)
Aluminium

MDL: MFCD00134029


Purity: 99.99% CAS No.: 7429-90-5 Physical Form: ≈0.0053g/m, Temper: as drawn Odor: Odorless MDL No.: MFCD00134029

Formula: Al
Boiling Pt: 2327 °C (1013 hPa)
Melting Pt: 660 °C
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR