Zirconium(IV) oxide ≥99.4% (metals basis excluding Hf) stabilized, powder -100+325 Mesh

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with calcia (CaO) 4%

40453-18 40453-A1
CAAA40453-18EA 70.47 CAD
CAAA40453-18 CAAA40453-30 CAAA40453-A1
Zirconium(IV) oxide ≥99.4% (metals basis excluding Hf) stabilized, powder -100+325 Mesh
Zirconium (IV) oxide

The main use of zirconia is in the production of ceramics. Zirconium dioxide is also used as the solid electrolyte in electrochromic devices As protective coating on particles of titanium dioxide pigments, as a refractory material, in insulation, abrasives and enamels.


Plasma Spray Grade. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, Alcohols, Halogens.

Formula: ZrO₂
Boiling Pt: ∼ 5000 °C
Melting Pt: ∼ 2700 °C
Density: 5.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR