Cobalt naphthenate ∼53% (6% Co) in mineral spirits

Supplier: Thermo Scientific Chemicals
Danger

40387-36 40387-A3
CAAA40387-36EA 251.22 CAD
CAAA40387-36 CAAA40387-A3
Cobalt naphthenate ∼53% (6% Co) in mineral spirits
Cobalt naphthenate

CAS No.: 61789-51-3 Flash Point: 40°(104°F) Physical Form: Dark Liquid UN No.: 1993 MDL No.: MFCD02094763

Formula: C₂₂H₁₄CoO₄
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD02094763
CAS Number: 61789-51-3
EINECS: 263-064-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR