Silicon carbide, ɑ-phase α-phase ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

40155-22 40155-36 40155-A3
CAAA40155-22EA 117.08 CAD
CAAA40155-22 CAAA40155-36 CAAA40155-A3
Silicon carbide, ɑ-phase α-phase ≥99.8% (metals basis)
Silicon carbide, α-phase

MDL: MFCD00049531


Purity: 99.8% CAS No.: 409-21-2 Molecular Formula: SiC Formula Weight: 40.10 Physical Form: 2 Micron Powder, Surface Area 9-11m2/g Odor: Odorless Merck Reference: 14,8492 MDL No.: MFCD00049531

Formula: SiC
Melting Pt: 2700 °C
Density: 3.22 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049531
CAS Number: 409-21-2
EINECS: 206-991-8
Merck Index: 14,8492

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR