Erbium(III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

40092-06 40092-09 40092-18
CAAA40092-06EA 77.1 CAD
CAAA40092-06 CAAA40092-09 CAAA40092-18
Erbium(III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis)
Erbium (III) sulphate octahydrate

Purity: 99.9% CAS No.: 10031-52-4 Molecular Formula: Er2(SO4)3.8H2O Formula Weight: 766.82 (622.70anhy) Physical Form: Crystalline Density: 3.205 Melting Point: dec. MDL No.: MFCD00149696

Formula: Er₂O₁₂S₃·8H₂O
Melting Pt: Dec.
Density: 3,205
Storage Temperature: Ambient
MDL Number: MFCD00149696
CAS Number: 10031-52-4
EINECS: 236-779-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR