Platinum 0,3% on alumina, pellets 2.7-3.3 mm (0.11-0.13 in), reduced

Supplier: Thermo Scientific Chemicals

Synonyms: Platinium

39826-14
CAAA39826-14EA 1229.24 CAD
CAAA39826-14
Platinum 0,3% on alumina, pellets 2.7-3.3 mm (0.11-0.13 in), reduced
Platinum

Uses: Selective hydrogenation, gas purifications: e.g. Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide


CAS No.: 7440-06-4 MDL No.: MFCD00011179


Applications: Selective hydrogenation, gas purifications: e.g. Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide

Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 215-691-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR