Vanadium(IV) etioporphyrin III oxide

Supplier: Thermo Scientific Chemicals

39586-MF
CAAA39586-MFEA 832.66 CAD
CAAA39586-MF
Vanadium(IV) etioporphyrin III oxide
Vanadium(IV) etioporphyrin III oxide

CAS No.: 15709-03-2 Molecular Formula: C32H36N4OV Formula Weight: 543.61 Physical Form: Crystalline Appearance: Purple Odor: Odorless MDL No.: MFCD00058901

Formula: C₃₂H₃₆N₄OV
MW: 543.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058901
CAS Number: 15709-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR