Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ytterbium(III) 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)ytterbium(III), Yb(TMHD)3

39581-03 39581-06
CAAA39581-03EA 125.77 CAD
CAAA39581-03 CAAA39581-06
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ytterbium(III) 99%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')ytterbium(III)

Purity: 99% CAS No.: 15492-52-1 Molecular Formula: C33H57O6Yb Formula Weight: 722.86 Physical Form: Crystalline Sensitivity: Hygroscopic MDL No.: MFCD00000034

Formula: C₃₃H₅₇O₆Yb
Melting Pt: 167…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00000034
CAS Number: 15492-52-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR