Lithium metagallate ≥99%

Supplier: Thermo Scientific Chemicals

39370-06
CAAA39370-06EA 176.96 CAD
CAAA39370-06 CAAA39370-14
Lithium metagallate ≥99%
Lithium metagallate

Purity: 99% CAS No.: 944251-30-3 Molecular Formula: GaLiO2 Formula Weight: 108.66 Physical Form: Powder Appearance: White Melting Point: 400° MDL No.: MFCD00210623

Formula: GaLiO₂
Melting Pt: >400 °C
Storage Temperature: Ambient
MDL Number: MFCD00210623
CAS Number: 944251-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR