Lithium niobium ethoxide 5% w/v ethanol , 99% Metals basis

Supplier: Thermo Scientific Chemicals
Danger

39155-AC 39155-AE
CAAA39155-ACEA 170.64 CAD
CAAA39155-AC CAAA39155-AE
Lithium niobium ethoxide 5% w/v ethanol , 99% Metals basis
Lithium niobium ethoxide

Purity: 99+% CAS No.: 86745-52-0 Molecular Formula: LiNb(OC2H5)6 Formula Weight: 370.21 Flash Point: 17°(63°F) Physical Form: Liquid Sensitivity: Moisture Sensitive Appearance: Yellow Odor: Alcohol-like UN No.: 1170 MDL No.: MFCD00210625

Formula: LiNb(OC₂H₅)₆
MW: 370.21 g/mol
Flash Pt: 17 °C (63 °F)
Storage Temperature: Ambient
MDL Number: MFCD00210625
CAS Number: 86745-52-0
UN: 1170
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR