Dichloromethane ≥99.5% stabilized ACS

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with amylene

39116-AP 39116-K2 39116-K4 39116-K7 39116-M1
CAAA39116-APEA 62.88 CAD
CAAA39116-AP CAAA39116-K2 CAAA39116-K4 CAAA39116-K7 CAAA39116-M1
Dichloromethane ≥99.5% stabilized ACS
Dichloromethane

Stabilized with amylene


Purity: 99.5+% CAS No.: 75-09-2 Molecular Formula: CH2Cl2 Formula Weight: 84.93 Physical Form: Liquid Density: 1.325 Refractive Index: 1.4244 Appearance: Colorless Odor: Sweet Melting Point: -95° Boiling Point: 39-40° Beilstein Reference: 1730800 Merck Reference: 14,6063 UN No.: 1593 MDL No.: MFCD00000881

Formula: CH₂Cl₂
Boiling Pt: 39…40 °C
Melting Pt: –95 °C
Density: 1.325
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 14,6063
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10
Evaporation residue 0.00002
Titratable acid 0.0003meq/g
Free halogens Pass test
H2O 0.0002


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR