Molybdenum, Plasma Standard Solution, Specpure®, Mo 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

38719-AD 38719-AE
CAAA38719-ADEA 84.85 CAD
CAAA38719-AD CAAA38719-AE CAAA38719-AP
Molybdenum, Plasma Standard Solution, Specpure®, Mo 1000 µg/ml, Thermo Scientific
Standards Cationic Standards Molybdenum Standards

Molecular Formula: (NH4)2MoO4 in 1% NH4OH
Physical Form: Liquid
Odor: Ammonia-like
MDL No.: MFCD00003465

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR