Phenolphthalein 0.5% (w/v) in alcohol

Supplier: Thermo Scientific Chemicals
Danger

38703-AP
CAAA38703-APEA 178.54 CAD
CAAA38703-AP
Phenolphthalein 0.5% (w/v) in alcohol
Phenolphthalein

MDL: MFCD00005913 Notes: Transition interval: pH 8.0 (colorless) to pH 10.0 (red)


CAS No.: 77-09-8 Molecular Formula: C20H14O4 Formula Weight: 318.33 Flash Point: 11°(52°F) Physical Form: Liquid Density: 0.918 Appearance: Colorless Odor: Alcohol-like UN No.: 1987 MDL No.: MFCD00005913

Formula: C₂₀H₁₄O₄
Density: 0.918
Flash Pt: 11 °C (52 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
UN: 1987
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR