Methyl orange 0.1% (w/v) in aqueous solution

Supplier: Thermo Scientific Chemicals

38695-AP
CAAA38695-APEA 57.51 CAD
CAAA38695-AP
Methyl orange 0.1% (w/v) in aqueous solution
Methyl orange

MDL: MFCD00007502 Notes: Transition interval: pH 3.2 (pink or red) to pH 4.4 (yellow)


CAS No.: 547-58-0 Molecular Formula: C14H14N3NaO3S Formula Weight: 327.34 Physical Form: Liquid Density: 0.987 Odor: Odorless MDL No.: MFCD00007502

Formula: C₁₄H₁₄N₃NaO₃S
Density: 0.987
Storage Temperature: Ambient
MDL Number: MFCD00007502
CAS Number: 547-58-0
EINECS: 208-925-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR