Palladium 5% on activated carbon powder wetted_50%_Water, eggshell, unreduced Type A401102-5

Supplier: Thermo Scientific Chemicals

38299-06 38299-14 38299-22
CAAA38299-06EA 130.67 CAD
CAAA38299-06 CAAA38299-14 CAAA38299-22
Palladium 5% on activated carbon powder wetted_50%_Water, eggshell, unreduced Type A401102-5
Palladium

Applications: Aromatic nitro group hydrogenation


CAS No.: 7440-05-3 Physical Form: 900-1100m2/g Appearance: Black Odor: Odorless MDL No.: MFCD03457879


Applications: Aromatic nitro group hydrogenation

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR