Aluminum calcium isopropoxide 10% (w/v) in isopropanol (IPA) , 99.8% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

36594-AC 36594-AE
CAAA36594-ACEA 145.36 CAD
CAAA36594-AC CAAA36594-AE
Aluminum calcium isopropoxide 10% (w/v) in isopropanol (IPA) , 99.8% (metals basis)
Aluminum calcium isopropoxide

Purity: 99.8% CAS No.: 23275-27-6 Molecular Formula: Ca[Al(OCH(CH3)2)4]2 Formula Weight: 566.30 Physical Form: Liquid Sensitivity: Moisture Sensitive Odor: Alcohol-like Boiling Point: 230-240°/4.0mm UN No.: 1219 MDL No.: MFCD00143586

Formula: Ca[Al(OCH(CH₃)₂)₄]₂
MW: 566.30 g/mol
Boiling Pt: 230…240 °C (4.0 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00143586
CAS Number: 23275-27-6
UN: 1219
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR