Gallic acid monohydrate 98+% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4,5-Trihydroxybenzoic acid monohydrate, 3,4,5-Trihydroxybenzoic acid hydrate

36417-22 36417-36
CAAA36417-22EA 106.02 CAD
CAAA36417-22 CAAA36417-36
Gallic acid monohydrate 98+% ACS
Gallic acid monohydrate

MDL: MFCD00149098


Purity: 98+% CAS No.: 5995-86-8 Molecular Formula: C7H6O5.H2O Formula Weight: 188.14 (170.12anhy) Physical Form: Powder Sensitivity: Light Sensitive Appearance: White to cream Melting Point: 250° dec. Merck Reference: 14,4345 MDL No.: MFCD00149098

Formula: C₇H₆O₅·1H₂O
MW: 188.14 g/mol
Melting Pt: 250 °C (dec.)
Flash Pt: 250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00149098
CAS Number: 5995-86-8
EINECS: 205-749-9
Merck Index: 14,4345

Order Now

Specification Test Results

Insoluble matter 0.01%
Residue after ignition 0.05%
SO3 0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR