Magnesium acetate tetrahydrate 98.0-102.0% ACS

Supplier: Thermo Scientific Chemicals

36358-22 36358-36 36358-A4
CAAA36358-22EA 56.09 CAD
CAAA36358-22 CAAA36358-36 CAAA36358-A4
Magnesium acetate tetrahydrate 98.0-102.0% ACS
Magnesium acetate tetrahydrate

MDL: MFCD00149214 Beilstein Registry No.: 3730605


Purity: 98.0-102.0% CAS No.: 16674-78-5 Molecular Formula: Mg(CH3COO)2.4H2O Formula Weight: 214.45 (142.39anhy) Physical Form: Crystalline Density: 1.454 Melting Point: 80° Beilstein Reference: 3730605 Merck Reference: 14,5654 MDL No.: MFCD00149214

Formula: (H₃CCOO)₂Mg·4H₂O
Melting Pt: 80 °C
Density: 1.454
Storage Temperature: Ambient
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 14,5654

Order Now

Specification Test Results

Insoluble matter 0.005% max.
Cl 0.001% max.
SO3 0.005% max.
Ba 0.001% max.
Ca 0.01% max.
Heavy Metals (as Pb) 5 ppm max.
Fe 5 ppm max.
Mn 0.001% max
K 0.005% max
NA 0.005% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR