Thulium(III) oxide ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

36279-04 36279-09
CAAA36279-04EA 206.98 CAD
CAAA36279-04 CAAA36279-09
Thulium(III) oxide ≥99.9% (REO, rare earth oxide basis)
Thulium (III) oxide

MDL: MFCD00011285 Notes: For vacuum deposition


Purity: 99.9% CAS No.: 12036-44-1 Molecular Formula: O3Tm2 Formula Weight: 385.87 Physical Form: 3-12mm Sintered Lump Appearance: Pale green Melting Point: 2425° Merck Reference: 14,9394 MDL No.: MFCD00011285

Formula: O₃Tm₂
Melting Pt: 2425 °C
Density: 8.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011285
CAS Number: 12036-44-1
EINECS: 234-851-6
Merck Index: 14,9394

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR