Chromium(III) oxide ≥99.6% (metals basis), fused lump 3-6mm

Supplier: Thermo Scientific Chemicals
Warning

36258-14 36258-22
CAAA36258-14EA 90.69 CAD
CAAA36258-14 CAAA36258-22
Chromium(III) oxide ≥99.6% (metals basis), fused lump 3-6mm
Chromium (III) oxide

For vacuum deposition


It can be used in selective oxidation. It is mainly used for smelting metal chromium and chromium carbide. It is used as coloring agent for color painting on enamelware and porcelain ware, imitation leather or building material. It is also used for making sun-proof coating, abrasive, green polishing compound and special printing ink for printing bank note. It is used as catalyst for organic synthetic chromium.


For vacuum deposition

Formula: Cr₂O₃
Boiling Pt: 3000 °C
Melting Pt: 2435 °C
Density: 5.22
Storage Temperature: Ambient
MDL Number: MFCD00010949
CAS Number: 1308-38-9
EINECS: 215-160-9
Merck Index: 14,2234

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR