Selenium ≥99.999% (metals basis), powder -200 mesh

Supplier: Thermo Scientific Chemicals
Danger

36208-18 36208-30
CAAA36208-18EA 395 CAD
CAAA36208-18 CAAA36208-30
Selenium ≥99.999% (metals basis), powder -200 mesh
Selenium

MDL: MFCD00134090


Purity: 99.999% CAS No.: 7782-49-2 UN No.: 3077 MDL No.: MFCD00134090

Formula: Se
Boiling Pt: ∼685 °C (1013 hPa)
Melting Pt: 217 °C
Density: 4.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134090
CAS Number: 7782-49-2
EINECS: 231-957-4
UN: 3288
ADR: 6.1,III
Merck Index: 12,08572

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR