Sodium borohydride 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

35788-22 35788-36
CAAA35788-22EA 145.36 CAD
CAAA35788-22 CAAA35788-36
Sodium borohydride 97+%
Sodium borohydride

As a reducing agent for organic, inorganic and organometallic compounds, as a bleaching agent, in the recycling of gold and platinum metals, in organic synthesis, nonaqueous reducing agent for aldehydes, ketones, and schiff bases


Purity: 97+% CAS No.: 16940-66-2 Molecular Formula: NaBH4 Formula Weight: 37.83 Physical Form: -10+40 Mesh Granules Density: 1.074 Appearance: White Odor: Weak Melting Point: 400° dec. Solubility: Soluble in water, ammonia and amines. Insoluble in ether Merck Reference: 14,8592 UN No.: 1426 MDL No.: MFCD00003518


As a reducing agent for organic, inorganic and organometallic compounds, as a bleaching agent, in the recycling of gold and platinum metals, in organic synthesis, nonaqueous reducing agent for aldehydes, ketones, and schiff bases

Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400 (dec.)
Density: 1.074
Flash Pt: 69
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3 I
Merck Index: 14,8592

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR