Gallium(II) chloride, ultra dry ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

35696-03
CAAA35696-03EA 111.55 CAD
CAAA35696-03
Gallium(II) chloride, ultra dry ≥99.999% (metals basis)
Gallium (II) chloride

Purity: 99.999% CAS No.: 24597-12-4 Molecular Formula: Ga2Cl4 Formula Weight: 281.26 Physical Form: Powder, Ampouled under argon Sensitivity: Air and Moisture Sensitive Appearance: White Odor: Acidic Melting Point: 164° Boiling Point: 535° UN No.: 3260 MDL No.: MFCD00151526

Formula: Ga₂Cl₄
MW: 281.26
Boiling Pt: 535 °C
Melting Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00151526
CAS Number: 13498-12-9
EINECS: 246-338-4
UN: 3260
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR