Cesium chloride, dry_ultra 99,998 Metals basis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cesium Chloride

35690-06 35690-14
CAAA35690-06EA 144.25 CAD
CAAA35690-06 CAAA35690-14
Cesium chloride, dry_ultra 99,998 Metals basis
Cesium chloride

MDL: MFCD00010955


Purity: 99.998% CAS No.: 7647-17-8 Molecular Formula: CsCl Formula Weight: 168.36 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.99 Refractive Index: 1.6418 Sensitivity: Hygroscopic Odor: Odorless Melting Point: 646° Boiling Point: 1303° Merck Reference: 14,2011 MDL No.: MFCD00010955

Formula: CsCl
MW: 168.36 g/mol
Boiling Pt: 1303
Melting Pt: 646
Density: 3.99
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2
Merck Index: 14,2011

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR