Bismuth(III) iodide, ultra dry ≥99.998% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bismuth triiodide

35681-06 35681-14
CAAA35681-06EA 259.12 CAD
CAAA35681-06 CAAA35681-14
Bismuth(III) iodide, ultra dry ≥99.998% (metals basis)
Bismuth (III) iodide

MDL: MFCD00010894


Purity: 99.998% CAS No.: 7787-64-6 Molecular Formula: BiI3 Formula Weight: 589.69 Physical Form: -10 Mesh Granules, Ampouled under argon Density: 5.778 Melting Point: 408° Boiling Point: ca 500° Merck Reference: 14,1269 UN No.: 3260 MDL No.: MFCD00010894

Formula: BiI₃
MW: 589.69 g/mol
Boiling Pt: ∼500 °C
Melting Pt: 408
Density: 5.778
Storage Temperature: Ambient
MDL Number: MFCD00010894
CAS Number: 7787-64-6
EINECS: 232-127-4
UN: 3260
ADR: 8 III
Merck Index: 14,1269

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR