Silicon ≥99.999% (metals basis), crystalline powder -325 mesh

Supplier: Thermo Scientific Chemicals
Warning

35662-09 35662-18
CAAA35662-09EA 143.15 CAD
CAAA35662-09 CAAA35662-18
Silicon ≥99.999% (metals basis), crystalline powder -325 mesh
Silicon

MDL: MFCD00085311


Purity: 99.999% CAS No.: 7440-21-3 Appearance: White Odor: Odorless UN No.: 1346 MDL No.: MFCD00085311

Formula: Si
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1,III
Merck Index: 12,08635

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR