Osmium(III) chloride trihydrate ≥99.99% (metals basis) Os 52-56%, Premion®

Supplier: Thermo Scientific Chemicals
Danger

33480-01 33480-03 33480-06
CAAA33480-01EA 150.57 CAD
CAAA33480-01 CAAA33480-03 CAAA33480-06
Osmium(III) chloride trihydrate ≥99.99% (metals basis) Os 52-56%, Premion®
Osmium (III) chloride trihydrate

Purity: 99.99% CAS No.: 135296-80-9 Molecular Formula: Cl3Os.3H2O Formula Weight: 350.60 (296.56anhy) Physical Form: Crystalline Appearance: Red-black Melting Point: dec. UN No.: 3260 MDL No.: MFCD00011148

Formula: Cl₃Os·H₂O
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00011148
CAS Number: 135296-80-9
EINECS: 236-587-7
UN: 3260
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR