Ceramic Tubes, AL23, Both Ends Open, Thermo Scientific
Supplier: Thermo Scientific Chemicals
31554-DR 31554-DW 31554-EH 31554-EM 31558-DM 31558-DW 31558-EO 31560-EM 31562-DW 31562-EO 31563-DR 31563-DW 31563-EH 31563-EM 31563-EO 031554-QAT 031556-QAT 031560-QAT 031562-QAT 031563-QAT
CAAA31553-DMEA 39.06 CAD
CAAA31553-DM CAAA31553-DR CAAA31553-DW CAAA31553-EH CAAA31553-EM CAAA31553-EO CAAA31554-DM CAAA31554-DR CAAA31554-DW CAAA31554-EH CAAA31554-EM CAAA31554-EO CAAA31555-DM CAAA31555-DR CAAA31555-DW CAAA31555-EH CAAA31555-EM CAAA31555-EO CAAA31556-DM CAAA31556-DR CAAA31556-DW CAAA31556-EH CAAA31556-EM CAAA31556-EO CAAA31558-DM CAAA31558-DR CAAA31558-DW CAAA31558-EH CAAA31558-EM CAAA31558-EO CAAA31560-DM CAAA31560-DR CAAA31560-DW CAAA31560-EH CAAA31560-EM CAAA31560-EO CAAA31562-DM CAAA31562-DR CAAA31562-DW CAAA31562-EH CAAA31562-EM CAAA31562-EO CAAA31563-DM CAAA31563-DR CAAA31563-DW CAAA31563-EH CAAA31563-EM CAAA31563-EO AA31553-QAT AA31554-QAT AA31555-QAT AA31556-QAT AA31558-QAT AA31560-QAT AA31562-QAT AA31563-QAT CAAA31553-QAT CAAA31554-QAT CAAA31555-QAT CAAA31556-QAT CAAA31558-QAT CAAA31560-QAT CAAA31562-QAT CAAA31563-QAT
Ceramic Tubes, AL23, Both Ends Open, Thermo Scientific
Tubing Transfer Tubing
Pure Al₂O₃ Aluminium oxide AL23.
  • Corrosion resistant
  • Permeable to microwaves
Dense, excellent thermal and electrical resistance properties.
Outer diameter ranges from 3 to 11 mm.