Ferroin (1,10-phenanthrolineferrous sulfate complex) 0.025 M in aqueous solution

Supplier: Thermo Scientific Chemicals

30562-18 30562-30
CAAA30562-18EA 69.52 CAD
CAAA30562-18 CAAA30562-30
Ferroin (1,10-phenanthrolineferrous sulfate complex) 0.025 M in aqueous solution
Ferroin (1,10-Phenanthroline-ferrous sulphate-complex)

MDL: MFCD00036428


CAS No.: 14634-91-4 Molecular Formula: C36H24FeN6O4S Formula Weight: 692.53 Physical Form: Liquid Appearance: Red

Formula: C₃₆H₂₄FeN₆SO₄
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036428
CAS Number: 14634-91-4
EINECS: 238-676-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR