1,3-Propylenediphosphonic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Propylenediphosphonic acid

30277-02 30277-04 30277-09
CAAA30277-02EA 48.51 CAD
CAAA30277-02 CAAA30277-04 CAAA30277-09
1,3-Propylenediphosphonic acid 98+%
1,3-Propylenediphosphonic acid

Purity: 98+% CAS No.: 4671-82-3 Molecular Formula: [P(O)(OH)2CH2]2CH2 Formula Weight: 204.06 Physical Form: Powder Odor: Odorless MDL No.: MFCD00050066

Formula: C₃H₁₀O₆P₂
MW: 204.06 g/mol
Melting Pt: 178 °C
Storage Temperature: Ambient
MDL Number: MFCD00050066
CAS Number: 4671-82-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR