Methyl ethyl ketone ≥99.5% for HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

22924-K2 22924-K7 22924-M1
CAAA22924-K2EA 122.45 CAD
CAAA22924-K2 CAAA22924-K7 CAAA22924-M1
Methyl ethyl ketone ≥99.5% for HPLC
Methyl ethyl ketone

MDL: MFCD00011648 Beilstein Registry No.: 741880 Fieser: 1,678


Purity: 99.5+% CAS No.: 78-93-3 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: -3°(26°F) Physical Form: Liquid Density: 0.805 Refractive Index: 1.3790 Appearance: Colorless Odor: Characteristic Melting Point: -87° Boiling Point: 80° Beilstein Reference: 741880 Merck Reference: 14,6072 UN No.: 1193 MDL No.: MFCD00011648

Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 80 °C
Melting Pt: –87 °C
Density: 0.805
Flash Pt: –3 °C (26 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3 II
REACH: 01-2119457290-43
Merck Index: 14,6072
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 15
Residue after evaporation 0.25%
Titratable acid 0.0005meq/g
H2O 0.2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR