N,N-Dimethylformamide 99,7 HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Formic acid dimethylamide, DMFA, Formyldimethylamine, N-Formyldimethylamine, Dimethylformamide, NSC 5356, DMF

22915-K2 22915-K7 22915-M1 22915-M6 22915-Y0
CAAA22915-K2EA 146.94 CAD
CAAA22915-K2 CAAA22915-K7 CAAA22915-M1 CAAA22915-M6 CAAA22915-Y0
N,N-Dimethylformamide 99,7 HPLC
N,N-Dimethylformamide

MDL: MFCD00003284 Beilstein Registry No.: 605365 Notes: Filtered through 0.2µ filters. Fieser: 1,273 9,182 11,198 12,203 14,148 16,144 18,146 19,137 21,178


Purity: 99.7+% CAS No.: 68-12-2 Molecular Formula: HCON(CH3)2 Formula Weight: 73.09 Flash Point: 57°(135°F) Physical Form: Liquid Density: 0.944 Refractive Index: 1.4310 Odor: Amine-like pH: 7 Melting Point: -61° Boiling Point: 153° Beilstein Reference: 605365 Merck Reference: 14,3243 UN No.: 2265 MDL No.: MFCD00003284

Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153
Melting Pt: -61
Density: 0.944
Flash Pt: 57 °C (135 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3 III
REACH: 01-2119475605-32
Merck Index: 14,3243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Evaporation residue 5 ppm
H2O 0.0005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR