Ammonium tetrathiotungstate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ammonium tungsten sulfide, Ammonium tetrathiotungstate(VI), Ammonium tungsten sulphide

22373-04 22373-09
CAAA22373-04EA 118.5 CAD
CAAA22373-04 CAAA22373-09
Ammonium tetrathiotungstate
Ammonium tetrathiotungstate

CAS No.: 13862-78-7 Molecular Formula: (NH4)2WS4 Formula Weight: 348.07 Physical Form: Chunks/Powder Sensitivity: Hygroscopic Odor: Like rotten eggs MDL No.: MFCD00061410

Formula: H₈N₂S₄W
MW: 348.158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061410
CAS Number: 13862-78-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR