Tetrabutylammonium dihydrogen phosphate 99+%

Supplier: Thermo Scientific Chemicals
Warning

19261-02 19261-04 19261-09 19261-18
CAAA19261-02EA 75.84 CAD
CAAA19261-02 CAAA19261-04 CAAA19261-09 CAAA19261-18
Tetrabutylammonium dihydrogen phosphate 99+%
Tetrabutylammonium dihydrogen phosphate

Used as a counter-ion in ion-pair chromatography of water-soluble vitamins
Soluble in acetonitrile, ethanol, water


Purity: 99+% CAS No.: 5574-97-0 Molecular Formula: C16H38NO4P Formula Weight: 339.46 Physical Form: Crystalline Appearance: White to off-white Melting Point: 151-154° Solubility: Soluble in acetonitrile, ethanol, water Beilstein Reference: 5196532 MDL No.: MFCD00064526


Used as a counter-ion in ion-pair chromatography of water-soluble vitamins

Formula: C₁₆H₃₈NO₄P
Melting Pt: 151…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00064526
CAS Number: 5574-97-0
EINECS: 226-947-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR