Gold(I) iodide 99%

Supplier: Thermo Scientific Chemicals
Danger

16617-03 16617-06
CAAA16617-03EA 322.32 CAD
CAAA16617-03 CAAA16617-06
Gold(I) iodide 99%
Gold (I) iodide

MDL: MFCD00014175 Insoluble in water. Soluble in alkali iodide, cyanide solutions. Decomposes in warm acids.


Purity: 99% CAS No.: 10294-31-2 Molecular Formula: AuI Formula Weight: 323.87 Physical Form: Crystalline Density: 8.25 Melting Point: 120° dec. Solubility: Insoluble in water. Soluble in alkali iodide, cyanide solutions. Decomposes in warm acids UN No.: 3260 MDL No.: MFCD00014175

Formula: AuI
Melting Pt: 120 °C (dec.)
Density: 8.25
Storage Temperature: Ambient
MDL Number: MFCD00014175
CAS Number: 10294-31-2
EINECS: 233-656-3
UN: 3260
ADR: 8,III
Merck Index: 12,04530

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR