Trimethylurea

Supplier: Thermo Scientific Chemicals

Synonyms: 1,1,3-Trimethylurea

16475-03 16475-06 16475-14
CAAA16475-03EA 46.45 CAD
CAAA16475-03 CAAA16475-06 CAAA16475-14
Trimethylurea
Trimethylurea

CAS No.: 632-14-4 Molecular Formula: (CH3)2NC(O)NH(CH3) Formula Weight: 102.14 Physical Form: Solid/semi-solid mass Sensitivity: Hygroscopic Odor: Characteristic MDL No.: MFCD00014868

Formula: C₄H₁₀N₂O
MW: 102.14 g/mol
Melting Pt: 74…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00014868
CAS Number: 632-14-4
EINECS: 211-169-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR