Barium silicon oxide

Supplier: Thermo Scientific Chemicals
Warning

15122-36
CAAA15122-36EA 366.56 CAD
CAAA15122-36
Barium silicon oxide
Barium silicon oxide

CAS No.: 13255-26-0 Molecular Formula: BaO and SiO2 Formula Weight: 213.40 Physical Form: Powder Density: 4.399 Refractive Index: 1.67 Melting Point: 1604° UN No.: 1564 MDL No.: MFCD00014186

Formula: BaO and SiO₂
Melting Pt: 1604 °C
Density: 4,399
Storage Temperature: Ambient
MDL Number: MFCD00014186
CAS Number: 13255-26-0
UN: 1564
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR