Barium carbonate 99,95 Metals basis

Supplier: Thermo Scientific Chemicals
Warning

14452-09 14452-18 14452-30 14452-A1
CAAA14452-09EA 70.31 CAD
CAAA14452-09 CAAA14452-18 CAAA14452-30 CAAA14452-A1
Barium carbonate 99,95 Metals basis
Barium carbonate

MDL: MFCD00003448 Notes: m.p. 174°C (90atm), 811°C (1 atm)


Purity: 99.95% CAS No.: 513-77-9 Molecular Formula: BaCO3 Formula Weight: 197.35 Physical Form: Lump/Powder Density: 4.43 Melting Point: 811° Boiling Point: dec. Merck Reference: 14,969 MDL No.: MFCD00003448

Formula: BaCO₃
MW: 197.34 g/mol
Boiling Pt: Dec.
Melting Pt: 1350
Density: 4.43
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1 III
Merck Index: 14,969

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR