Barium, plasma standard solution, Specpure®, Ba 10,000µg/ml, Thermo Scientific

Supplier: Thermo Scientific Chemicals

14423-AD 14423-AE 14423-AP
CAAA14423-ADEA 122.92 CAD
CAAA14423-AD CAAA14423-AE CAAA14423-AP
Barium, plasma standard solution, Specpure®, Ba 10,000µg/ml, Thermo Scientific
Standards Cationic Standards Barium Standards

Molecular Formula: Ba(NO3)2 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134031

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR