Lead(II) nitrate ≥99.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

14243-36 14243-A3
CAAAA14243-36EA 115.02 CAD
CAAAA14243-36 CAAA14243-A3
Lead(II) nitrate ≥99.0% ACS
Lead (II) nitrate

As an oxidizer in dye industry, in manufacturing of mother-of-pearl, as mordant in printing textile, as sensitizer in photography


Purity: 99.0% min CAS No.: 10099-74-8 Molecular Formula: Pb(NO3)2 Formula Weight: 331.20 Physical Form: Crystalline Density: 4.530 Appearance: White Odor: Odorless Melting Point: 470° dec. Solubility: Freely soluble in water. Soluble in absolute alcohol, methanol. Insoluble in concentrated HNO3 Merck Reference: 14,5414 UN No.: 1469 MDL No.: MFCD00011153


As an oxidizer in dye industry, in manufacturing of mother-of-pearl, as mordant in printing textile, as sensitizer in photography

Formula: Pb(NO₃)₂
Melting Pt: 470 °C (dec.)
Density: 4.530
Storage Temperature: Ambient
MDL Number: MFCD00011153
CAS Number: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1,II
Merck Index: 14,5414

Order Now

Specification Test Results

Form Crystalline
Cl 0.1%
Ca 0.005%
Cu 0.002%
Fe 0.001%
K 0.005%
NA 0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR